Please login to your online account to display your discounted pricing

Sodium Borate, 10-Hydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$405.80 - $500.45

Chemical Identifiers

Chemical Name or Material Sodium Borate
CAS 1303-96-4
Name Note 10-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003998

 
avantor performance
357005
Poly Bottle 2.5kg Each for $500.45

02003999

 
avantor performance
357001
Poly Bottle 500g Each for $405.80
Case of 4 Each for $1,623.20
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 381.37
Name Note 10-Hydrate
Packaging Poly Bottle
Physical Form Powder
Specific Gravity 1L = 1.73kg
Molecular Formula Na2B4O7·10H2O
CAS 1303-96-4
Quantity 2.5kg
Chemical Name or Material Sodium Borate