Please login to your online account to display your discounted pricing

Sodium Borate, 10-Hydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  357001

 View more versions of this product

Catalog No. 02003999

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 381.37
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Sodium Borate
Molecular Formula Na2B4O7·10H2O
Physical Form Powder
CAS 1303-96-4
Specific Gravity 1L = 1.73kg
Packaging Poly Bottle
Name Note 10-Hydrate
Quantity 500g

Specifications

Chemical Name or Material Sodium Borate
CAS 1303-96-4
Name Note 10-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37