Please login to your online account to display your discounted pricing

Sodium Borate, 10-Hydrate, N.F., J.T.Baker™

$125.58 - $592.12

Chemical Identifiers

Chemical Name or Material Sodium Borate
CAS 1303-96-4
Name Note 10-Hydrate
Purity Grade NF
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003996

 
avantor performance
357405
Poly Bottle 2.5kg Each for $592.12

02003997

 
avantor performance
357401
Poly Bottle 500g Each for $125.58
Description & Specifications

Specifications

Formula Weight 381.37
Chemical Name or Material Sodium Borate
CAS 1303-96-4
Molecular Formula Na2B4O7·10H2O
Quantity 2.5kg
Name Note 10-Hydrate
Purity Grade NF
Specific Gravity 1L = 1.73kg
Packaging Poly Bottle