Please login to your online account to display your discounted pricing

Sodium Borate, 10-Hydrate, N.F., J.T.Baker™


Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  357401

 View more versions of this product

Catalog No. 02003997

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 381.37
Chemical Name or Material Sodium Borate
Molecular Formula Na2B4O7·10H2O
Name Note 10-Hydrate
Packaging Poly Bottle
Purity Grade NF
Specific Gravity 1L = 1.73kg
CAS 1303-96-4
Quantity 500g

Specifications

Chemical Name or Material Sodium Borate
CAS 1303-96-4
Name Note 10-Hydrate
Purity Grade NF
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37