Please login to your online account to display your discounted pricing

Sodium Borate, 10-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$425.56 - $425.56

Chemical Identifiers

Chemical Name or Material Sodium Borate
CAS 1303-96-4
Name Note 10-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004001

 
avantor performance
356805
Poly Bottle 2.5kg Each for $425.56
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 381.37
Physical Form Crystals
Chemical Name or Material Sodium Borate
Packaging Poly Bottle
Name Note 10-Hydrate
Specific Gravity 1L = 1.73kg
Quantity 2.5kg
Molecular Formula Na2B4O7·10H2O
CAS 1303-96-4