Please login to your online account to display your discounted pricing

Sodium Borate, 10-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  3568-01

 View all options for this product

Catalog No. 02004002

 

Description & Specifications

Chemical Identifiers

Packaging Poly Bottle
Quantity 500g
Specific Gravity 1L = 1.73kg
Quantity 500g
Chemical Name or Material Sodium Borate
Name Note 10-Hydrate
CAS 1303-96-4
Purity Grade BAKER ANALYZED™ ACS Reagent
Physical Form Crystal
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37g/mol

Specifications

Chemical Name or Material Sodium Borate
CAS 1303-96-4
Name Note 10-Hydrate
Purity Grade BAKER ANALYZED™ ACS Reagent
Molecular Formula Na2B4O7·10H2O
Formula Weight 381.37g/mol