Please login to your online account to display your discounted pricing

Sodium Bisulfite FCC, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging STAKMOR

Manufacturer: avantor performance  744420

 View more versions of this product

Catalog No. MK744412

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging STAKMOR
Quantity 12kg
Chemical Name or Material Sodium Bisulfite
Name Note Purified
CAS 7631-90-5
Purity Grade FCC
Molecular Formula NaHSO3
Formula Weight 104.06g/mol

Specifications

Chemical Name or Material Sodium Bisulfite
Name Note Purified
CAS 7631-90-5
Purity Grade FCC
Molecular Formula NaHSO3
Formula Weight 104.06g/mol