Please login to your online account to display your discounted pricing

Sodium Bisulfite FCC, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Bisulfite
Name Note Purified
CAS 7631-90-5
Purity Grade FCC Grade
Molecular Formula NaHSO3
Formula Weight 104.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7444212

 
avantor performance
744406
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium Bisulfite
Name Note Purified
CAS 7631-90-5
Purity Grade FCC Grade
Molecular Formula NaHSO3
Formula Weight 104.06
Packaging Poly
Quantity 2.5kg