Please login to your online account to display your discounted pricing

Sodium Bisulfate, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5kg
Packaging Poly Bottle

Manufacturer: avantor performance  353405

 View more versions of this product

Catalog No. 02003992

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 138.07
Chemical Name or Material Sodium Bisulfate
Molecular Formula NaHSO4·H2O
Name Note Monohydrate
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 2.10kg
CAS 10034-88-5
Quantity 2.5kg

Specifications

Chemical Name or Material Sodium Bisulfate
CAS 10034-88-5
Name Note Monohydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaHSO4·H2O
Formula Weight 138.07