Please login to your online account to display your discounted pricing

Sodium Bisulfate, Monohydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$267.45 - $804.82

Chemical Identifiers

Chemical Name or Material Sodium Bisulfate
CAS 10034-88-5
Name Note Monohydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaHSO4·H2O
Formula Weight 138.07
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003993

 
avantor performance
353401
Poly Bottle 500g Each for $267.45

02003992

 
avantor performance
353405
Poly Bottle 2.5kg Each for $804.82
Description & Specifications

Specifications

Formula Weight 138.07
Physical Form Crystals
CAS 10034-88-5
Chemical Name or Material Sodium Bisulfate
Molecular Formula NaHSO4·H2O
Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Purity Grade BAKER ANALYZED™ Reagent
Name Note Monohydrate
Quantity 500g