missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium bisulfate monohydrate, 99%, for analysis, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC125000100
Specifications
Sodium bisulfate monohydrate | |
99% | |
NaHSO4·H2O | |
15,8722 | |
Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and,h2so4 | |
O.OS(=O)(=O)[O-].[Na+] | |
138.08 | |
138.08 | |
Analysis | |
Plastic drum | |
Solubility in water: 670g/L. Other solubilities: decomposes by alcohol into sodium sulfate and, h2so4 | |
10kg |
10034-88-5 | |
HNaO4S·H2O | |
0.001% max | |
3kls9y77yj, acmc-20ajox, sodium bisulfate monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate monohydrate, sodium hydrogen sulfate-1-hydrate, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, hydrate, sulfuric acid, monosodium salt, monohydrate, unii-3kls9y77yj | |
JXHZRQHZVYDRGX-UHFFFAOYSA-M | |
sodium;hydrogen sulfate;hydrate | |
23673662 | |
99% | |
0.0005% max | |
0.001% max | |
58.0°C |
Chemical Identifiers
10034-88-5 | |
138.08 | |
3kls9y77yj, acmc-20ajox, sodium bisulfate monohydrate, sodium hydrate hydrogen sulfate, sodium hydrogen sulfate monohydrate, sodium hydrogen sulfate-1-hydrate, sodium hydrogensulfate hydrate, sulfuric acid, monosodium salt, hydrate, sulfuric acid, monosodium salt, monohydrate, unii-3kls9y77yj | |
sodium;hydrogen sulfate;hydrate |
HNaO4S·H2O | |
JXHZRQHZVYDRGX-UHFFFAOYSA-M | |
23673662 | |
O.OS(=O)(=O)[O-].[Na+] |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger