missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Bisulfate AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 10034-88-5
Physical Form Crystalline
Molecular Formula NaHSO4·H2O
Formula Weight 138.08
Chemical Name or Material Sodium Bisulfate, Monohydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK7432500 Avantor Performance
743212
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10034-88-5
NaHSO4·H2O
Sodium Bisulfate, Monohydrate
Crystalline
138.08

Specifications

10034-88-5
NaHSO4·H2O
POLYSTORMOR
Sodium Bisulfate, Monohydrate
Crystalline
138.08
500g