missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium bis(trimethylsilyl)amide, 1M soln. in THF, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L08779AP

 View more versions of this product

Catalog No. AAL08779AP



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bis(trimethylsilyl)amide is useful for the generation of carbenes, the amination of sulfonamides, and for the preparation of lanthanide complexes. It is significantly accelerated the polymerization of pheneylacetylene in conjunction with rhodium (I) catalysis. It is used to deprotonate ketones and esters to generate enolate derivatives, phosphonium salts, generating Wittig reagents.

Solubility
Soluble in benzene, toluene and ether. Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents, Oxygen. Store away from water and oxidizing agents.
Specifications

Specifications

Sodium bis(trimethylsilyl)amide
500 mL
MFCD00009835
3629917
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
WRIKHQLVHPKCJU-UHFFFAOYSA-N
sodium;bis(trimethylsilyl)azanide
2724254
1M soln. in THF
0.904
1070-89-9
C6H18NNaSi2
UN2924
−17°C (1°F)
Soluble in benzene,toluene and ether. Reacts with water.
C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
183.377
183.38
Moisture sensitive; May form a precipitate

Chemical Identifiers

1070-89-9
183.377
WRIKHQLVHPKCJU-UHFFFAOYSA-N
2724254
C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
C6H18NNaSi2
MFCD00009835
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
sodium;bis(trimethylsilyl)azanide
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335-H336-H351

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 213-983-8

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only