missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 4664006

 View more versions of this product

Catalog No. AA4664006



Specifications

Specifications

Sodium bis (trifluoromethylsulfonyl)imide
257°C to 258°C
91742-21-1
(CF3SO2)2NNa
UN3261
QXZNUMVOKMLCEX-UHFFFAOYSA-N
sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
131887538
Solid
Moisture Sensitive
5g
C2HF6NNaO4S2
MFCD14156021
Sodium triflimide; Sodium trifluoromethanesulfonimide
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]
304.133
303.13

Chemical Identifiers

91742-21-1
304.133
QXZNUMVOKMLCEX-UHFFFAOYSA-N
131887538
C(F)(F)(F)S(=O)(=O)NS(=O)(=O)C(F)(F)F.[Na]
C2HF6NNaO4S2
MFCD14156021
Sodium triflimide; Sodium trifluoromethanesulfonimide
sodium;1,1,1-trifluoro-N-(trifluoromethylsulfonyl)methanesulfonamide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

SDS Documents