missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Bis(fluorosulfonyl)imide, TCI America™

Manufacturer:  TCI America S09925G

 View more versions of this product

Catalog No. S09925G



Specifications

Specifications

Sodium Bis(fluorosulfonyl)imide
112°C
100669-96-3
1759
[N-](S(=O)(=O)F)S(=O)(=O)F.[Na+]
203.11
203.11
≥98.0% (T)
White
5g
F2NNaO4S2
VCCATSJUUVERFU-UHFFFAOYSA-N
sodium;bis(fluorosulfonyl)azanide
102310776
Crystal-Powder at 20°C

Chemical Identifiers

100669-96-3
203.11
102310776
[N-](S(=O)(=O)F)S(=O)(=O)F.[Na+]
F2NNaO4S2
VCCATSJUUVERFU-UHFFFAOYSA-N
sodium;bis(fluorosulfonyl)azanide
Safety and Handling

Safety and Handling

TSCA : No

Documents