missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium bis(2-methoxyethoxy)aluminium hydride, 70 wt% sol. in toluene (ca. 3.5M), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 187900010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagentSpecifications
Sodium bis(2-methoxyethoxy)aluminium hydride | |
C6H16AlNaO4 | |
02,382; 03,260; 04,441; 05,596; 06,528; 07,327; 08,448; 12,440; 15,290 | |
Solubility in water: reacts. Other solubilities: soluble in aromatic hydrocarbons and ethers | |
68.5 to 71.5% | |
1.036 | |
1kg |
22722-98-1,108-88-3 | |
[(CH3OCH2CH2O)2AlH2]Na | |
1.0360g/mL | |
202.16 | |
Glass bottle | |
65 mPa.s (20°C) | |
4°C |
Safety and Handling
Danger
RUO â Research Use Only