missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium biphenyl, 20%w/w solution in diethylene glycol diethyl ether, offered as 20X, ACROS Organics™

$236.95 - $236.95

Chemical Identifiers

CAS 5137-46-2,112-36-7
Molecular Formula C12H10Na
Linear Formula C6H5C6H5·Na
Formula Weight 177.2
MDL Number MFCD00013182
Synonym Biphenylyl sodium, solution in diethylene glycol diethyl ether
Chemical Name or Material Sodium biphenyl, 20%w/w solution in diethylene glycol diethyl ether, 20%w/w Solution in Diethylene Glycol Diethyl Ether
Assay 17 to 23% (w/w) (typically), actual strength on certificate of analysis
Grade Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC419423000 SDS Thermo Fisher Scientific
419423000
300mL Each for $236.95
Specifications

Description

Reduction reagent
Description & Specifications

Chemical Identifiers

5137-46-2,112-36-7
C6H5C6H5·Na
MFCD00013182
Sodium biphenyl, 20%w/w solution in diethylene glycol diethyl ether, 20%w/w Solution in Diethylene Glycol Diethyl Ether
Reagent Grade
C12H10Na
177.2
Biphenylyl sodium, solution in diethylene glycol diethyl ether
17 to 23% (w/w) (typically), actual strength on certificate of analysis

Specifications

177.2
300mL
1.0030g/mL
Biphenylyl sodium, solution in diethylene glycol diethyl ether
C12H10Na
MFCD00013182
17 to 23% (w/w) (typically), actual strength on certificate of analysis
Solution
Dark green to blue-black
112-36-7,5137-46-2
Reagent may partially crystallize
C6H5C6H5·Na
Sodium biphenyl, 20%w/w solution in diethylene glycol diethyl ether, 20%w/w Solution in Diethylene Glycol Diethyl Ether
Reagent Grade

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 225-894-1