Please login to your online account to display your discounted pricing

Sodium Bicarbonate USP GenAR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Bicarbonate
CAS 144-55-8
Purity Grade USP
Physical Form Powder
Molecular Formula NaHCO3
Formula Weight 84.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK7749212

 
avantor performance
774906
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 84.01
Quantity 2.5kg
Purity Grade USP
Chemical Name or Material Sodium Bicarbonate
Packaging Poly
Physical Form Powder
Molecular Formula NaHCO3
CAS 144-55-8