Please login to your online account to display your discounted pricing

Sodium Bicarbonate-Sodium Chloride Solution, LabChem™

$51.69 - $51.69

Chemical Identifiers

Chemical Name or Material Sodium Bicarbonate-Sodium Chloride Solution
CAS 7732-18-5
CAS Max % 99.33
CAS 7647-14-5
CAS Max % 0.59
CAS 144-55-8
CAS Max % 0.08
Linear Formula NaHCO3
Formula Weight 84.01
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC229482

 
lab chem
LC229482
1L Each for $51.69
Description & Specifications

Specifications

Formula Weight 84.01
Identification Passes Test
Solubility Soluble in water
CAS 7732-18-5
Quantity 1L
CAS Max % 99.33
Chemical Name or Material Sodium Bicarbonate-Sodium Chloride Solution
CAS Max % 0.08
Packaging Poly Bottle
Physical Form Liquid
Storage Information Room Temperature
Linear Formula NaHCO3
Decomposition Information Hydrogen chloride
Color Clear, colorless
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7647-14-5
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.59
CAS 144-55-8
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.