Please login to your online account to display your discounted pricing

Sodium Bicarbonate, For Chlorine Dioxide, 5.5% (w/v), LabChem™

$44.37 - $44.37

Chemical Identifiers

Chemical Name or Material Sodium Bicarbonate
CAS 7732-18-5
CAS Max % 94.5
CAS 144-55-8
CAS Max % 5.5
Name Note For Chlorine Dioxide
Linear Formula NaHCO3
Formula Weight 84.01
Synonym Baking Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC229451

 
lab chem
LC229451
500mL Each for $44.37
Description & Specifications

Specifications

Formula Weight 84.01
Chemical Name or Material Sodium Bicarbonate
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
Name Note For Chlorine Dioxide
Physical Form Liquid
Assay 5.5% (w/v)
Linear Formula NaHCO3
Density 1g/mL
CAS 7732-18-5
Synonym Baking Soda
CAS Max % 94.5
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 144-55-8
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 5.5
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Refrigerate
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.