Learn More
Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP), Fisher Chemical™
Manufacturer: Fisher Chemical S637212
Specifications
Sodium Bicarbonate | |
White | |
8.3 | |
1 EU/mg max. (USP) | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
84.01 | |
99.0 to 101.0% (EP) | |
5ppm max. (JP) | |
Pass Test (FCC) | |
2mg/kg max. (FCC) | |
0.004% max. (USP) | |
0.002% max. (USP) | |
20ppm max. (BP) | |
Meets Requirements (BP) | |
Pass Test (BP) | |
150ppm max. (BP) | |
1 I.U./mg max. (EP) | |
Pass Test (BP) | |
Clear and Colorless (JP) | |
>50°C |
144-55-8 | |
270°C | |
2.5kg | |
CHNaO3 | |
acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogen carbonate, sodium hydrogencarbonate | |
C(=O)(O)[O-].[Na+] | |
84.006 | |
CHEBI:32139 | |
Powder/Solid | |
BP/EP/FCC/JP/USP | |
Pass Test (BP) | |
20ppm max. (BP) | |
0.25% max. (FCC) | |
2μg/g max. (USP) | |
Pass Test (USP) | |
Poly Bottle | |
Pass Test | |
2μg/g max. (USP) | |
0.015% max. (USP) | |
0.01% max. (USP) | |
0.015% max. (USP) | |
1μg/g max. (USP) |
Chemical Identifiers
144-55-8 | |
84.006 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
sodium;hydrogen carbonate |
CHNaO3 | |
MFCD00003528 | |
acidosan, baking soda, bicarbonate of soda, carbonic acid monosodium salt, meylon, neut, sodium acid carbonate, sodium bicarbonate, sodium hydrogen carbonate, sodium hydrogencarbonate | |
CHEBI:32139 | |
C(=O)(O)[O-].[Na+] |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. May cause adverse kidney effects. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:1