missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Bicarbonate, ACS Grade, 99.7 to 100.3%, LabChem™

$52.11 - $52.11

Chemical Identifiers

CAS 144-55-8
CAS Max % 100
Linear Formula NaHCO3
Formula Weight 84.01
Synonym Baking Soda
Chemical Name or Material Sodium Bicarbonate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC229431 Lab Chem
LC229431
500g Each for $52.11
Specifications Description & Specifications

Chemical Identifiers

144-55-8
NaHCO3
Baking Soda
ACS Grade
100
84.01
Sodium Bicarbonate

Specifications

144-55-8
Powder
84.01
2.159g/cm3
Poly Bottle
2.159g/cm3
Carbon monoxide; Carbon dioxide
270°C
Sodium Bicarbonate
ACS Grade
100
NaHCO3
Baking Soda
Passes Test
Soluble in water
Room Temperature
White
500g
99.7 to 100.3%

Safety and Handling

GHS H Statement
Causes eye irritation.

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 205-633-8