Please login to your online account to display your discounted pricing

Sodium Bicarbonate ACS, EMD Millipore


Quantity 12kg
Packaging Poly Pail

Manufacturer: emd millipore corporation  SX03205

 View more versions of this product

Catalog No. MSX03205

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 84.1
Purity Grade ACS/EP/Reagent Grade
Quantity 12kg
Bulk Density 1000kg/m3
Density 2.22g/cm3 (20°C)
Chemical Name or Material Sodium Bicarbonate
Name Note EMSURE™, For Analysis
Packaging Poly Pail
Molecular Formula CHNaO3
Linear Formula NaHCO3
CAS 144-55-8
Vapor Pressure 8.3 hPa (30°C)
Synonym Sodium bicarbonate
Recommended Storage Store without restrictions

Specifications

Chemical Name or Material Sodium Bicarbonate
Name Note EMSURE™, For Analysis
CAS 144-55-8
Purity Grade ACS/EP/Reagent Grade
Molecular Formula CHNaO3
Linear Formula NaHCO3
Formula Weight 84.1
Synonym Sodium bicarbonate
Accessories

Store without restrictions