missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Bicarbonate ACS, EMD Millipore

Manufacturer:  Emd Millipore Corporation SX03203

 View more versions of this product

Catalog No. MSX03203


 

Specifications Description & Specifications

Specifications

ACS/EP/Reagent Grade
2.5kg
2.22g/cm3 (20°C)
Poly Bottle
Sodium bicarbonate
EMSURE™, For Analysis
Sodium Bicarbonate
84.1
1000kg/m3
144-55-8
8.3 hPa (30°C)
CHNaO3
NaHCO3

Chemical Identifiers

144-55-8
NaHCO3
Sodium bicarbonate
ACS/EP/Reagent Grade
CHNaO3
84.1
Sodium Bicarbonate, EMSURE™, For Analysis

Safety and Handling

Recommended Storage : Store without restrictions