missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Bicarbonate ACS, EMD Millipore

$41.77 - $272.37

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Linear Formula NaHCO3
Formula Weight 84.1
Synonym Sodium bicarbonate
Chemical Name or Material Sodium Bicarbonate, EMSURE™, For Analysis
Grade ACS/EP/Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MSX03201 Emd Millipore Corporation
SX03201
Poly Bottle 500g Each for $41.77
MSX03203 Emd Millipore Corporation
SX03203
Poly Bottle 2.5kg Each for $141.89
MSX03205 Emd Millipore Corporation
SX03205
Poly Pail 12kg Each for $272.37
Specifications Description & Specifications

Chemical Identifiers

144-55-8
NaHCO3
Sodium bicarbonate
ACS/EP/Reagent Grade
CHNaO3
84.1
Sodium Bicarbonate, EMSURE™, For Analysis

Specifications

144-55-8
NaHCO3
Sodium bicarbonate
2.22g/cm3 (20°C)
8.3 hPa (30°C)
Sodium Bicarbonate, EMSURE™, For Analysis
CHNaO3
84.1
1000kg/m3
Poly Bottle
500g
ACS/EP/Reagent Grade

Safety and Handling

Recommended Storage : Store without restrictions