missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Bicarbonate ACS AR, Macron Fine Chemicals™
macron
$1512.67 - $1512.67
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
Synonym | sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
IUPAC Name | sodium hydrogen carbonate |
SMILES | [Na+].OC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
MK7412500
|
Avantor
741212 |
500 g | POLYSTORMOR |
Case of 4 Each for $1,512.67
|
|
||||
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
sodium hydrogen carbonate |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
CHEBI:32139 | |
[Na+].OC([O-])=O |
Specifications
144-55-8 | |
CHNaO3 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
ACS | |
Sodium Bicarbonate |
500 g | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium hydrogen carbonate | |
516892 | |
84.01 | |
POLYSTORMOR |