missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Benzohydroxamate Hydrate, TCI America™

$210.84 - $565.30

Chemical Identifiers

CAS 22513-32-2
Molecular Formula C7H8NNaO3
Molecular Weight (g/mol) 177.135
MDL Number MFCD00058967
InChI Key CTXNFHAZPJAZQN-UHFFFAOYSA-N
Synonym Benzohydroxamic Acid Sodium Salt
PubChem CID 56924513
IUPAC Name sodium;N-oxidobenzamide;hydrate
SMILES C1=CC=C(C=C1)C(=O)N[O-].O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B01125G View Documents TCI America
B01125G
5g Each for $210.84
 
B011225G View Documents TCI America
B011225G
25g Each for $565.30
 
Specifications

Chemical Identifiers

22513-32-2
177.135
CTXNFHAZPJAZQN-UHFFFAOYSA-N
56924513
C1=CC=C(C=C1)C(=O)N[O-].O.[Na+]
C7H8NNaO3
MFCD00058967
Benzohydroxamic Acid Sodium Salt
sodium;N-oxidobenzamide;hydrate

Specifications

White-Yellow
22513-32-2
MFCD00058967
CTXNFHAZPJAZQN-UHFFFAOYSA-N
sodium;N-oxidobenzamide;hydrate
56924513
Crystal-Powder at 20°C
Sodium Benzohydroxamate Hydrate
5g
C7H8NNaO3
Benzohydroxamic Acid Sodium Salt
C1=CC=C(C=C1)C(=O)N[O-].O.[Na+]
177.135
159.12
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No