Please login to your online account to display your discounted pricing

Sodium Benzoate, N.F. - F.C.C., J.T.Baker™

$558.55 - $558.55

Chemical Identifiers

Chemical Name or Material Sodium Benzoate
CAS 532-32-1
Purity Grade FCC/NF
Molecular Formula C6H5COONa
Formula Weight 144.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003982

 
avantor performance
350005
Poly Bottle 2.5kg Each for $558.55
Description & Specifications

Specifications

Purity Grade FCC/NF
Formula Weight 144.11
Packaging Poly Bottle
Specific Gravity 1L = 1.44kg
Molecular Formula C6H5COONa
CAS 532-32-1
Quantity 2.5kg
Chemical Name or Material Sodium Benzoate