missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Benzoate (Granular/NF/FCC), Fisher Chemical

Manufacturer:  Fisher Chemical S224500

Catalog No. S224-500




Description

Description

Specifications

Specifications

0.04% max. (FCC), Pass Test (USP)
Pass Test (FCC/USP)
Pass Test
White
500g
532-32-1
C6H5CO2Na
antimol, benzoate of soda, benzoate sodium, benzoate, sodium, benzoic acid sodium salt, benzoic acid, sodium salt, caswell no. 746, natrium benzoicum, sobenate, sodium benzoate
[Na+].[O-]C(=O)C1=CC=CC=C1
144.11
144.11
99.0 to 100.5% (USP)
Sodium Benzoate
Glass Bottle
1.5% max. (FCC/USP)
300°C
8
C7H5NaO2
MFCD00012463
WXMKPNITSTVMEF-UHFFFAOYSA-M
sodium benzoate
517055
Powder/Solid
FCC/NF

Chemical Identifiers

532-32-1
144.11
WXMKPNITSTVMEF-UHFFFAOYSA-M
517055
[Na+].[O-]C(=O)C1=CC=CC=C1
C7H5NaO2
MFCD00012463
antimol, benzoate of soda, benzoate sodium, benzoate, sodium, benzoic acid sodium salt, benzoic acid, sodium salt, caswell no. 746, natrium benzoicum, sobenate, sodium benzoate
sodium benzoate
Safety and Handling

Safety and Handling

SDS Documents