missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Azide, White Powder, Fisher BioReagents

Manufacturer:  Fisher BioReagents FLBP922I500

Catalog No. BP922I-500



Description

Description

  • Sodium Azide is readily soluble in water
  • Handle with caution and follow all storage recommendations
  • For research use only
Specifications

Specifications

Sodium Azide
N3Na
azide, sodium, azoture de sodium, hydrazoic acid, sodium salt, kazoe, natriumazid, natriumazid german, nemazyd, smite, sodium azide, sodium azide na n3
[N-]=[N+]=[N-].[Na+]
65.011
CHEBI:278547
Powder/Solid
Poly Bottle/Poison Pack
White
10
26628-22-8
15, 8717
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium;azide
33557
65.01
1.850g/cm³
300°C
275°C
500g

Chemical Identifiers

26628-22-8
65.011
azide, sodium, azoture de sodium, hydrazoic acid, sodium salt, kazoe, natriumazid, natriumazid german, nemazyd, smite, sodium azide, sodium azide na n3
CHEBI:278547
[N-]=[N+]=[N-].[Na+]
N3Na
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
33557
sodium;azide
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents
Product Certifications
Promotions

Promotions