Please login to your online account to display your discounted pricing

Sodium Azide, Practical grade, 97% min., EMD Millipore


Quantity 100g

Manufacturer: emd millipore corporation  SX03001

 View more versions of this product

Catalog No. MSX03001

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Practical Grade
Formula Weight 65.1
Quantity 100g
Color Light yellow to off-white
Chemical Name or Material Sodium Azide
Physical Form Crystalline
Assay Percent Range 97%, min.
Molecular Formula NaN3
Linear Formula NaN3
CAS 26628-22-8
Synonym Sodium trinitride
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Sodium Azide
CAS 26628-22-8
Purity Grade Practical Grade
Physical Form Crystalline
Assay Percent Range 97%, min.
Molecular Formula NaN3
Linear Formula NaN3
Formula Weight 65.1
Synonym Sodium trinitride
Accessories

Room Temperature