missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Azide, Practical grade, 97% min., EMD Millipore

$144.29 - $518.74

Chemical Identifiers

CAS 26628-22-8
Physical Form Crystalline
Assay Percent Range 97%, min.
Molecular Formula NaN3
Linear Formula NaN3
Formula Weight 65.1
Synonym Sodium trinitride
Chemical Name or Material Sodium Azide
Grade Practical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MSX03001 Emd Millipore Corporation
SX03001
100g Each for $144.29
MSX03003 Emd Millipore Corporation
SX03003
500g Each for $518.74
Specifications Description & Specifications

Chemical Identifiers

26628-22-8
97%, min.
NaN3
Sodium trinitride
Practical Grade
Crystalline
NaN3
65.1
Sodium Azide

Specifications

26628-22-8
97%, min.
NaN3
Sodium trinitride
100g
Practical Grade
Crystalline
NaN3
65.1
Light yellow to off-white
Sodium Azide

Safety and Handling

Recommended Storage : Room Temperature