Please login to your online account to display your discounted pricing

Sodium Azide OR, Macron Fine Chemicals™


macron

Quantity 100g
Packaging Poly

Manufacturer: avantor performance  195357

 View more versions of this product

Catalog No. MK1953100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 65.01
Quantity 100g
Chemical Name or Material Sodium Azide
Molecular Formula NaN3
CAS 26628-22-8
Packaging Poly

Specifications

Chemical Name or Material Sodium Azide
CAS 26628-22-8
Molecular Formula NaN3
Formula Weight 65.01