Please login to your online account to display your discounted pricing

Sodium Azide OR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Sodium Azide
CAS 26628-22-8
Molecular Formula NaN3
Formula Weight 65.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK1953100

 
avantor performance
195357
Poly 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK19535

 
avantor performance
195309
Poly Pail 5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK1953500

 
avantor performance
195359
Poly 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 65.01
Quantity 100g
Chemical Name or Material Sodium Azide
Molecular Formula NaN3
CAS 26628-22-8
Packaging Poly