Please login to your online account to display your discounted pricing

Sodium Azide, Crystalline, Laboratory, Fisher Chemical


Quantity 500g
Packaging Poly Bottle

Manufacturer: fisher scientific company  S227I500

 View more versions of this product

Catalog No. S227I-500

  • / Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 300°C
Purity Grade Laboratory Grade
Physical Form Powder/Solid
Formula Weight 65.01
Assay ≥99%
Melting Point 275°C
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Density 1.850g/cm³
Color White
pH 10
Identification Pass test
Assay Percent Range ≥99 %
Linear Formula NaN3
CAS 26628-22-8
ChemAlert Storage Symbol Blue
Chemical Name or Material Sodium Azide
MDL Number MFCD00003536
Molecular Formula N3Na
Synonym Sodium Trinitride
Packaging Poly Bottle
Health Hazard 1 Signal Word:DANGER!
Health Hazard 3 Emergency Overview
Very toxic if swallowed. Very toxic in contact with skin. May be fatal if absorbed through skin or swallowed. Toxic by inhalation. May cause pulmonary edema. Irritating to eyes, respiratory syste

Specifications

Chemical Name or Material Sodium Azide
CAS 26628-22-8
Purity Grade Laboratory Grade
Physical Form Powder/Solid
Assay ≥99%
Assay Percent Range ≥99 %
Molecular Formula N3Na
Linear Formula NaN3
Formula Weight 65.01
MDL Number MFCD00003536
Synonym Sodium Trinitride

  • Source of azide anion for organic chemical synthesis
  • Azide anions are strong nucleophiles and azide functionalized molecules can undergo multiple reactions
  • Sodium azide can also serve as preservative for aqueous solutions
  • Handle with caution
  • Storage recommendations should be followed
  • For research use only

Accessories
 

Signal Word:DANGER!

Emergency Overview
Very toxic if swallowed. Very toxic in contact with skin. May be fatal if absorbed through skin or swallowed. Toxic by inhalation. May cause pulmonary edema. Irritating to eyes, respiratory syste

SDS & Safety Documents