Learn More
Sodium azide, 99%, extra pure, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 190380025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
C-X bond formation reagent, For Azidation/Diazo transferSpecifications
Sodium azide | |
26628-22-8 | |
98.5% min | |
NaN3 | |
01,1041; 02,376; 03,259; 04,440; 05,593; 15,289; 17,68 | |
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
sodium;azide | |
33557 | |
65.01 | |
Extra Pure | |
Plastic bottle | |
>275°C |
99% | |
2.5kg | |
N3Na | |
MFCD00003536 | |
15, 8717 | |
Solubility in water: 420g/L (17°C). Other solubilities: soluble in liquid ammonia, slightly soluble in alcohol, insoluble in diethyl ether | |
[N-]=[N+]=[N-].[Na+] | |
65.01 | |
CHEBI:278547 | |
99% | |
0.5% max. | |
1.85 | |
1.8500g/mL |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Danger
EINECSNumber : 247-852-1
RTECSNumber : VY8050000
TSCA : TSCA
RUO â Research Use Only