Learn More
Sodium azide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01431436
Description
In airbag inflation, as a preservative in diagnostic medicinals.Sodium azide is used in organic synthesis for the introduction of an azide group to prepare a desired compound. It acts as a precursor to prepare inorganic azides like lead azide and silver azide. In biochemistry, it is useful as a probe reagent and a preservative. It also acts as a nitridizing agent and cytochrome oxidase inhibitor. Further, it is used as a propellant in airbags.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium Azide | |
26628-22-8 | |
500 g | |
MFCD00003536 | |
Soluble in water and liquid ammonia. Slightly soluble in alcohol. Insoluble in ether. | |
[N-]=[N+]=[N-].[Na+] | |
65.011 | |
CHEBI:278547 | |
Air Sensitive |
∽275° (decomposition) | |
Powder or Granules | |
N3Na | |
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
sodium;azide | |
33557 | |
65.01 | |
1.846 |
Chemical Identifiers
26628-22-8 | |
65.011 | |
PXIPVTKHYLBLMZ-UHFFFAOYSA-N | |
33557 | |
sodium;azide |
N3Na | |
MFCD00003536 | |
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german | |
CHEBI:278547 | |
[N-]=[N+]=[N-].[Na+] |
Safety and Handling
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
H300+H310+H330-H373
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 247-852-1
RTECSNumber : VY8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only