missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Azide, 99%, Alfa Aesar

Manufacturer:  Alfa Aesar 1431422

 View more versions of this product

Catalog No. AA1431422



Specifications

Specifications

Sodium Azide
26628-22-8
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
[N-]=[N+]=[N-].[Na+]
65.011
CHEBI:278547
100g
N3Na
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium;azide
33557
65.01

Chemical Identifiers

26628-22-8
65.011
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
CHEBI:278547
[N-]=[N+]=[N-].[Na+]
N3Na
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
33557
sodium;azide
Documents