Please login to your online account to display your discounted pricing

Sodium Azide, 99.0% min., EMD Millipore


Quantity 100g

Manufacturer: emd millipore corporation  SX02991

 View more versions of this product

Catalog No. MSX02991

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 65.1
Physical Form Crystals
Color White to off-white
Quantity 100g
Density 1.85g/cm3 (20°C)
Assay Percent Range 99.0% min.
Synonym Sodium trinitride
Name Note For Synthesis
CAS 26628-22-8
Recommended Storage Store at +15°C to +25°C
Chemical Name or Material Sodium Azide
Molecular Formula NaN3

Specifications

Chemical Name or Material Sodium Azide
Name Note For Synthesis
CAS 26628-22-8
Physical Form Crystals
Assay Percent Range 99.0% min.
Molecular Formula NaN3
Formula Weight 65.1
Synonym Sodium trinitride
Accessories

Store at +15°C to +25°C