Please login to your online account to display your discounted pricing

Sodium Azide, 99.0% min., EMD Millipore

$168.11 - $328.23

Chemical Identifiers

Chemical Name or Material Sodium Azide
Name Note For Synthesis
CAS 26628-22-8
Physical Form Crystals
Assay Percent Range 99.0% min.
Molecular Formula NaN3
Formula Weight 65.1
Synonym Sodium trinitride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MSX02991

 
emd millipore corporation
SX02991
100g Each for $168.11

MSX02993

 
emd millipore corporation
SX02993
500g Each for $328.23
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 65.1
Color White to off-white
Quantity 100g
Density 1.85g/cm3 (20°C)
Chemical Name or Material Sodium Azide
Name Note For Synthesis
Assay Percent Range 99.0% min.
Molecular Formula NaN3
CAS 26628-22-8
Synonym Sodium trinitride
Recommended Storage Store at +15°C to +25°C
Safety and Handling

Store at +15°C to +25°C