missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Azide, For Chromium, 0.5% (w/v), LabChem™

Manufacturer:  LabChem LC229402

 View more versions of this product

Catalog No. LC229402



Specifications

Specifications

Sodium Azide
26628-22-8,7732-18-5
N3Na
1g/mL
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
sodium;azide
33557
65.01
0.5% (w/v)
Poly Bottle
1g/mL
Nitrogen oxides
1L
For Chromium
99.5,0.5
NaN3
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
[N-]=[N+]=[N-].[Na+]
65.011
CHEBI:278547
Liquid
Passes Test
Soluble in water
Room Temperature
Colorless

Chemical Identifiers

26628-22-8
65.011
sodium azide, azide, sodium, natriumazid, nemazyd, smite, kazoe, azoture de sodium, sodium azide na n3, hydrazoic acid, sodium salt, natriumazid german
CHEBI:278547
[N-]=[N+]=[N-].[Na+]
N3Na
PXIPVTKHYLBLMZ-UHFFFAOYSA-N
33557
sodium;azide
Safety and Handling

Safety and Handling

GHS H Statement
Harmful to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

Documents
Promotions

Promotions