Learn More
Sodium aurothiomalate(I), 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03974006
Description
Sodium aurothiomalate(I) is useful for high mobility group box chromosomal protein 1 western blotting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium aurothiomalate(I) | |
Sulphurous | |
C4H5AuNa2O5S | |
5 g | |
gold sodium thiomalate | |
YLQOAPBVYJCTPW-UHFFFAOYNA-K | |
gold;sodium;2-sulfanylbutanedioic acid | |
133108869 | |
99.9% |
12244-57-4 | |
(metals basis) | |
MFCD00064304,MFCD00064304 | |
14,4518 | |
Very soluble in water; Practically insoluble in alcohol, ether | |
O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O | |
408.09 | |
390.08 | |
Powder |
Chemical Identifiers
12244-57-4 | |
408.09 | |
YLQOAPBVYJCTPW-UHFFFAOYNA-K | |
133108869 | |
O.[Na+].[Na+].[Au+].[O-]C(=O)CC([S-])C([O-])=O |
C4H5AuNa2O5S | |
MFCD00064304,MFCD00064304 | |
gold sodium thiomalate | |
gold;sodium;2-sulfanylbutanedioic acid |
Safety and Handling
GHS H Statement
H301-H332-H317
Toxic if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280g-P301+P312-P302+P352-P304+P340-P312-P330-P333+P313-P363-P501c
H302+H332-H317
EINECSNumber : 235-479-7
RTECSNumber : MD5435000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only