missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Ascorbate, USP, 99-101%, Spectrum™ Chemical

SO108, 134-03-2, C6H7NaO6

$4406.94 - $11476.51

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606311
View Documents Promotion Details
Spectrum Chemical Mfg Cor
SO1085KG
5 kg Poly Pail
Each for $4,406.94
 
18606312
View Documents Promotion Details
Spectrum Chemical Mfg Cor
SO10850KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Sodium Ascorbate, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

134-03-2
198.11
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
C6H7NaO6
IFVCRSPJFHGFCG-HXPAKLQESA-N
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

134-03-2
7.0 to 8.0
Poly Pail
5 kg
IFVCRSPJFHGFCG-HXPAKLQESA-N
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
99 to 101%
1
0.0025
C6H7NaO6
+103° to +108°
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
198.11
USP
SDS
missing translation for 'documents'

missing translation for 'documents'