missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Ascorbate, USP, 99-101%, Spectrum™ Chemical

SO108, 134-03-2, C6H7NaO6

$211.03 - $706.26

Chemical Identifiers

CAS 134-03-2
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
IUPAC Name sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606309
View Documents Promotion Details
Spectrum Chemical Mfg Cor
SO108100GM
100 g Amber Glass Bottle
Each for $211.03
 
18606310
View Documents Promotion Details
Spectrum Chemical Mfg Cor
SO108500GM
500 g Amber Glass Bottle
Each for $706.26
 
Description

Description

Spectrum™ Chemical Sodium Ascorbate, USP is used to prevent or treat low levels of vitamin C in people who do not get enough of the vitamin from their diets. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

134-03-2
198.11
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
C6H7NaO6
IFVCRSPJFHGFCG-HXPAKLQESA-N
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

134-03-2
7.0 to 8.0
Amber Glass Bottle
100 g
IFVCRSPJFHGFCG-HXPAKLQESA-N
sodium 5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide
99 to 101%
1
0.0025
C6H7NaO6
+103° to +108°
[Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O
198.11
USP
SDS
missing translation for 'documents'

missing translation for 'documents'