Please login to your online account to display your discounted pricing

Sodium Arsenite, Reagent Grade, LabChem™

$442.43 - $442.43

Chemical Identifiers

Chemical Name or Material Sodium Arsenite
CAS 7784-46-5
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula NaAsO2
Formula Weight 129.91
UN Number UN2027
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC228709

 
lab chem
LC228709
100g Each for $442.43
Description & Specifications

Specifications

Formula Weight 129.91
Health Hazard 1 Danger
UN Number UN2027
CAS Max % 100
CAS 7784-46-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Store locked up in a well-ventilated place.
Keep container tightly closed.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center or doctor.
If on skin: Wash with plenty of soap and water.
Immediately call a poison center/doctor.
Take off immediately all contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
Health Hazard 2 GHS H Statement
Fatal if swallowed or in contact with skin.
Toxic if inhaled.
May cause cancer (oral, inhalation).
Toxic to aquatic life with long lasting effects.
EINECS Number 232-070-5
Linear Formula NaAsO2
Packaging Poly Bottle
Physical Form Powder
Decomposition Information Arsenic and its oxides
Solubility Soluble in water
Quantity 100g
Chemical Name or Material Sodium Arsenite
Specific Gravity 1.87g/cm3
Identification Passes Test
Color White
Purity Grade Reagent Grade
Storage Information Room Temperature
Density 1.87g/cm3
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust.
Do not get in eyes, on skin, or on clothing.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
Store locked up in a well-ventilated place.
Keep container tightly closed.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Immediately call a poison center or doctor.
If on skin: Wash with plenty of soap and water.
Immediately call a poison center/doctor.
Take off immediately all contaminated clothing and wash it before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Fatal if swallowed or in contact with skin.
Toxic if inhaled.
May cause cancer (oral, inhalation).
Toxic to aquatic life with long lasting effects.