Please login to your online account to display your discounted pricing

Sodium Arsenite, For Residual Chlorine, Chlorine Dioxide, 0.5% (w/v), LabChem™

$88.26 - $88.26

Chemical Identifiers

Chemical Name or Material Sodium Arsenite
CAS 7732-18-5
CAS Max % 99.5
CAS 7784-46-5
CAS Max % 0.5
Name Note For Residual Chlorine, Chlorine Dioxide
Linear Formula NaAsO2
Formula Weight 129.91
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC228801

 
lab chem
LC228801
500mL Each for $88.26
Description & Specifications

Specifications

Formula Weight 129.91
Chemical Name or Material Sodium Arsenite
Decomposition Information Arsenic and its oxides
Color Clear, colorless
Quantity 500mL
Name Note For Residual Chlorine, Chlorine Dioxide
Physical Form Liquid
Assay 0.5% (w/v)
Linear Formula NaAsO2
Density 1g/mL
CAS 7732-18-5
Health Hazard 1 Danger
CAS Max % 99.5
Health Hazard 2 GHS H Statement
May cause cancer (oral, inhalation).
Identification Passes Test
CAS 7784-46-5
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, protective clothing, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 0.5
Solubility Soluble in water
Specific Gravity 1g/mL
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, protective clothing, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause cancer (oral, inhalation).