missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Arsenite, 0.100 Molar, Ricca Chemical

$66.97 - $100.48

Chemical Identifiers

CAS 7732-18-5,1310-73-2,1327-53-3,144-55-8,7647-01-0 (Hydrochloric Acid),7732-18-5 (Water, Deionized)
CAS Min % 95.0,1.55,1.03,0.38
CAS Max % 99.0,1.61,1.08,0.4
Physical Form Liquid
Chemical Name or Material Sodium Arsenite
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
7142-16 Ricca Chemical Company
RI714216
Glass amber 500mL Each for $66.97
7142-32 Ricca Chemical Company
RI714232
Glass amber 1L Each for $100.48
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,1310-73-2,1327-53-3,144-55-8,7647-01-0 (Hydrochloric Acid),7732-18-5 (Water, Deionized)
99.0,1.61,1.08,0.4
Sodium Arsenite
95.0,1.55,1.03,0.38
Liquid

Specifications

Liquid
500mL
95.0,1.55,1.03,0.38
7732-18-5 (Water, Deionized),144-55-8,7647-01-0 (Hydrochloric Acid),1327-53-3,7732-18-5,1310-73-2
Sodium Arsenite
Clear colorless
0.100Molar
99.0,1.61,0.4,1.08
Glass amber