Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium antimonate trihydrate, 98+%

Chemical Identifiers

Chemical Name or Material Sodium antimonate trihydrate
CAS 15432-85-6
Molecular Formula NaSbO3·3H2O
Formula Weight 246.80 (192.75 Anhydrous)
MDL Number MFCD00058775
UN Number UN1549
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1614118

 
alfa aesar™
A1614118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1614130

 
alfa aesar™
A1614130
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 246.80 (192.75 Anhydrous)
Chemical Name or Material Sodium antimonate trihydrate
TSCA TSCA
Odor Odorless
Assay ≥98%
Molecular Formula NaSbO3·3H2O
MDL Number MFCD00058775
CAS 15432-85-6
UN Number UN1549
Quantity 50g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.

Applications
A fine powder of sodium antimonate trihydrate, a slightly soluble salt of the highest available was used to prepare a dilute solution of sodium antimonate. In polyol method it is found that the ferrite is impossibly gained when diethylene glycol is replaced by ethylene glycol, or anhydrous sodium acetate by acetic acid sodium salt trihydrate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from incompatible materials such as strong oxidizing agents, strong acids, strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.