missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04032036

 View more versions of this product

Catalog No. AA4032036



Description

Description

Used in the laboratory as an essential ingredient of the microcosmic salt bead test for identification of metallic radicals. Also used as intermediates. Finds application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium ammonium hydrogen phosphate tetrahydrate
White
Crystals
H13NNaO8P
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
IQTQISLCLWJRPM-UHFFFAOYSA-M
azanium;sodium;hydrogen phosphate;tetrahydrate
56924540
Reagent
White crystals
79°C (decomposition)
7783-13-3
500 g
MFCD00066970
Soluble in water. Insoluble in alcohol
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]
209.067
209.07 (137.02 Anhydrous)
Odorless
1.544

Chemical Identifiers

7783-13-3
209.067
IQTQISLCLWJRPM-UHFFFAOYSA-M
56924540
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]
H13NNaO8P
MFCD00066970
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
azanium;sodium;hydrogen phosphate;tetrahydrate
Safety and Handling

Safety and Handling

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only