Please login to your online account to display your discounted pricing

Sodium aluminium hydride, 93%, ACROS Organics™


Quantity 100g

Manufacturer: thermo fisher scientific  AC380111000

 View more versions of this product

Catalog No. AC380111000

  • / Each
 

Description & Specifications

Chemical Identifiers

CAS 13770-96-2
Assay Percent Range 93%
Molecular Formula H4AlNa
Formula Weight 54
MDL Number MFCD00192443
Quantity 100g

Specifications

CAS 13770-96-2
Assay Percent Range 93%
Molecular Formula H4AlNa
Formula Weight 54
MDL Number MFCD00192443